Skip to Content
MilliporeSigma

Skip To

02400

Ethanolamine

ACS reagent grade, ≥99.0% (GC/NT), puriss. p.a.

Synonym(s):

2-Aminoethanol, 2-Aminoethyl alcohol, ETA, MEA, MEA 90, MEA-LCI, Monoethanolamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
NH2CH2CH2OH
CAS Number:
Molecular Weight:
61.08
NACRES:
NA.21
PubChem Substance ID:
UNSPSC Code:
12352100
EC Number:
205-483-3
MDL number:
Beilstein/REAXYS Number:
505944
Assay:
≥99.0% (GC/NT)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)

Quality Segment

vapor density

2.1 (vs air)

vapor pressure

0.2 mmHg ( 20 °C)

grade

ACS reagent, puriss. p.a.

assay

≥99.0% (GC/NT)

autoignition temp.

1436 °F

expl. lim.

17 %

impurities

≤0.2% water

ign. residue

≤0.05% (as SO4)

refractive index

n20/D 1.454

bp

170 °C (lit.), 69-70 °C/10 mmHg

mp

10-11 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

cation traces

Ca: ≤5 mg/kg, Cd: ≤1 mg/kg, Co: ≤1 mg/kg, Cr: ≤1 mg/kg, Cu: ≤1 mg/kg, Fe: ≤2 mg/kg, K: ≤20 mg/kg, Mg: ≤1 mg/kg, Mn: ≤1 mg/kg, Na: ≤20 mg/kg, Ni: ≤1 mg/kg, Pb: ≤1 mg/kg, Zn: ≤10 mg/kg

functional group

amine, hydroxyl

SMILES string

NCCO

InChI

1S/C2H7NO/c3-1-2-4/h4H,1-3H2

InChI key

HZAXFHJVJLSVMW-UHFFFAOYSA-N

General description

Ethanolamine is an alkanolamine. Ethanolamine is an essential growth-promoting supplement for serum-free medium for culturing mouse plasmacytomas and hybridoma cell lines of MPC11-BL and SP2/0-BL origins.[1] It is reported to efficiently inhibit the coupling reaction of aryl halides.

Application

Ethanolamine was used for the synthesis of N-oleoylaminoethanol standard.[2] It was also used for the preparation and derivatization of poly(acrylic acid) (PAA) films. It may be used in the scintillating method for evaluating counts for carbon-14.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
4110003981361.00844
assay

≥99.0% (GC/NT)

assay

≥99.5%

assay

≥99.0%

assay

98.0-100.5% (acidimetric)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

500

bp

170 °C (lit.), 69-70 °C/10 mmHg

bp

170 °C (lit.), 69-70 °C/10 mmHg

bp

170 °C (lit.), 69-70 °C/10 mmHg

bp

171 °C/1013 hPa

refractive index

n20/D 1.454

refractive index

n20/D 1.454 (lit.)

refractive index

n20/D 1.454 (lit.)

refractive index

-

density

1.012 g/mL at 25 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

density

1.02 g/cm3 at 20 °C

vapor density

2.1 (vs air)

vapor density

2.1 (vs air)

vapor density

2.1 (vs air)

vapor density

-


Still not finding the right product?

Explore all of our products under Ethanolamine


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 2

flash_point_f

195.8 °F - Pensky-Martens closed cup

flash_point_c

91 °C - Pensky-Martens closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What is the concentration of this solution?

    1 answer
    1. Product No. 02400 contains neat ethanolamine specified at ≥99.0% purity. Using the minimum specified purity, density, and molecular weight of ethanolamine, this neat product would have a concentration of about 16.4 M at 25 °C.

      Helpful?

Reviews

No rating value

Active Filters