Skip to Content
MilliporeSigma
All Photos(2)

Documents

01542

Supelco

Chrysophanol

analytical standard

Synonym(s):

1,8-Dihydroxy-3-methylanthraquinone, 3-Methylchrysazin, Chrysophanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O4
CAS Number:
Molecular Weight:
254.24
Beilstein:
1252300
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

192-196 °C

suitability

conforms to structure for Proton NMR spectrum

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

Cc1cc(O)c2C(=O)c3c(O)cccc3C(=O)c2c1

InChI

1S/C15H10O4/c1-7-5-9-13(11(17)6-7)15(19)12-8(14(9)18)3-2-4-10(12)16/h2-6,16-17H,1H3

InChI key

LQGUBLBATBMXHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chrysophanol is an anthraquinone compound and occurs naturally in Chinese herb Rheum officinale. It shows antimicrobial activity, and is an active component in many crude drugs having purgative properties.

Application

It may be used as reference standard in analyzing bioactive components (hyperin, nobiletin, hesperidin, emodin, chrysophanol, resveratrol, curcumine, and physcion) in Danning tablets using HPLC-UV and HPLC-ESI-MS.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: aloe

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chrysophanol, an antimicrobial anthraquinone from the root extract of Colubrina greggii.
Garcia-Sosa, Karlina, et al.
Journal of the Mexican Chemical Society, 50.2, 76-78 (2006)
Chrysophanol induces necrosis through the production of ROS and alteration of ATP levels in J5 human liver cancer cells.
Lu CC
Molecular Nutrition And Food Research, 54(7), 967-976 (2010)
Myung Sun Lee et al.
Phytotherapy research : PTR, 25(6), 833-837 (2010-11-23)
Inactivation of epidermal growth factor receptor (EGFR) is a prime method used in colon cancer therapy. Here it is shown that chrysophanic acid, a natural anthraquinone, has anticancer activity in EGFR-overexpressing SNU-C5 human colon cancer cells. Chrysophanic acid preferentially blocked
Simultaneous analysis of eight bioactive compounds in Danning tablet by HPLC-ESI-MS and HPLC-UV.
Liu R
Journal of Pharmaceutical and Biomedical Analysis, 43(3), 1007-1012 (2007)
Anne C Jones et al.
Die Naturwissenschaften, 99(7), 583-586 (2012-06-23)
The nonnative hemlock woolly adelgid (Adelges tsugae Annand, Hemiptera: Sternorrhyncha: Adelgidae) has been a significant mortality agent of eastern hemlock (Tsuga canadensis Carriere) throughout a large portion of its geographic range. During a study investigating adelgid vigor in relation to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service