Skip to Content
MilliporeSigma

PHL89757

Physcion

phyproof® Reference Substance

Synonym(s):

1,8-Dihydroxy-3-methoxy-6-methylanthraquinone, 6-O-Methylemodin, 6-Methoxychrysophanic acid, Emodin-3-methyl ether, Parietin, Rheochrysidin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
6770499
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

primary reference standard

product line

phyproof® Reference Substance

assay

≥90.0% (HPLC)

form

powder

manufacturer/tradename

PhytoLab

SMILES string

COc1cc(O)c2C(=O)c3c(O)cc(C)cc3C(=O)c2c1

InChI

1S/C16H12O5/c1-7-3-9-13(11(17)4-7)16(20)14-10(15(9)19)5-8(21-2)6-12(14)18/h3-6,17-18H,1-2H3

InChI key

FFWOKTFYGVYKIR-UHFFFAOYSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Anthraquinone-derivative found in medicinal herbs.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG


Still not finding the right product?

Explore all of our products under Physcion


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library