Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00239

Supelco

Micro particles, magnetic, carboxy functionalized

Synonym(s):

Magnetic micro particles, carboxy functionalized

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.25

grade

analytical standard

Quality Level

description

particle mass concentration 18 - 22 mg/mL

bp

212 °C

mp

32 °C

application(s)

glass & ceramic
industrial qc
pharmaceutical

format

neat

storage temp.

2-8°C

General description

Carboxy functionalized magnetic microparticles are a particle size standard, designed for profiling particle size distribution (PSD) of test samples.

Application

Used for equipment particle size calibration.
It can also be used to:
  • study biochemicals and reagents as well as magnetic microparticles.
  • better understand the mechanism by which toxic airborne particles can affect cardiovascular function.

Features and Benefits

  • suitable for routine instrument calibration checks, testing and corrections
  • available in 10 mL pack size as a neat sample

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Skin exposure to micro-and nano-particles can cause haemostasis in zebrafish larvae
McLeish JA, et al.
Thrombosis and Haemostasis, 103(04), 797-807 (2010)
Jennifer A McLeish et al.
Thrombosis and haemostasis, 103(4), 797-807 (2010-02-23)
Low mass ambient exposure to airborne particles is associated with atherothrombotic events that may be a consequence of the combustion-derived nanoparticle content. There is concern also over the potential cardiovascular impact of manufactured nanoparticles. To better understand the mechanism by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service