Skip to Content
MilliporeSigma
All Photos(1)

Documents

30-1940

Sigma-Aldrich

Thiourea

JIS special grade, ≥98.0%

Synonym(s):

Sulfourea, Thiocarbamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CSNH2
CAS Number:
Molecular Weight:
76.12
Beilstein:
605327
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

JIS special grade

Assay

≥98.0%

form

solid

availability

available only in Japan

mp

170-176 °C (lit.)

SMILES string

NC(N)=S

InChI

1S/CH4N2S/c2-1(3)4/h(H4,2,3,4)

InChI key

UMGDCJDMYOKAJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chaotropic agent; strong denaturant. Increases solubility and recovery of proteins

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mireille Vonlanthen et al.
The Journal of organic chemistry, 78(8), 3980-3988 (2013-03-09)
Proof that sulfur is a viable reporting element for the development of fluorescent chemosensors for metal ions is presented. To date, the majority of metal-responsive fluorescent chemosensors have relied on metal-nitrogen coordination to provide a fluorescence response, most commonly by
Mathieu P Lalonde et al.
Journal of the American Chemical Society, 135(5), 1891-1894 (2013-01-17)
A highly enantio- and diastereoselective synthesis of indolo- and benzoquinolizidine compounds has been developed through the formal aza-Diels-Alder reaction of enones with cyclic imines. This transformation is catalyzed by a new bifunctional primary aminothiourea that achieves simultaneous activation of both
Shrinivas Venkataraman et al.
Macromolecular rapid communications, 34(8), 652-658 (2013-03-14)
Readily water-soluble PEGylated amphiphiles containing bis-thiourea-based molecular recognition units at the interface of hydrophobic and hydrophilic blocks are developed. Self-assembly of these amphiphiles is found to be dependent on the exact chemical composition of the hydrophobic component. Elongated, spherical, and
Yoshiji Takemoto
Chemical & pharmaceutical bulletin, 58(5), 593-601 (2010-05-13)
We have developed several multifunctional thiourea catalysts bearing a tertiary amine or an 1,2-amino alcohol in expectation of their synchronous activation of a nucleophile and an electrophile through both acid-base and hydrogen-bonding interactions. From these studies, it was revealed that
Ai-Fang Li et al.
Chemical Society reviews, 39(10), 3729-3745 (2010-08-26)
This critical review highlights recent advances in the structurally modified (thio)urea-based receptors for anion complexation and sensing. Modifications of the (thio)urea structure are aimed at a better anion binding in terms of higher binding constant, anion selectivity and feasibility. Major

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service