MilliporeSigma
  • Anion complexation and sensing using modified urea and thiourea-based receptors.
Anion complexation and sensing using modified urea and thiourea-based receptors.
Chemical Society reviews (2010-08-26)
Ai-Fang Li, Jin-He Wang, Fang Wang, Yun-Bao Jiang
ABSTRACT
This critical review highlights recent advances in the structurally modified (thio)urea-based receptors for anion complexation and sensing. Modifications of the (thio)urea structure are aimed at a better anion binding in terms of higher binding constant, anion selectivity and feasibility. Major (thio)urea receptors are reviewed as N-alkyl, N-aryl and N-amido/N-amino (thio)ureas. Hints for designing (thio)urea-based receptors for anions are discussed (102 references).
Sigma-Aldrich
Thiourea, JIS special grade, ≥98.0%
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%