Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-5810

Sigma-Aldrich

5-Sulfosalicylic acid dihydrate

SAJ first grade, ≥99.0%

Synonym(s):

2-Hydroxy-5-sulfobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO3SC6H3-2-(OH)CO2H·2H2O
CAS Number:
Molecular Weight:
254.21
Beilstein:
650741
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ first grade

Assay

≥99.0%

form

solid

availability

available only in Japan

mp

105-110 °C (lit.)

SMILES string

[H]O[H].[H]O[H].OC(=O)c1cc(ccc1O)S(O)(=O)=O

InChI

1S/C7H6O6S.2H2O/c8-6-2-1-4(14(11,12)13)3-5(6)7(9)10;;/h1-3,8H,(H,9,10)(H,11,12,13);2*1H2

InChI key

BHDKTFQBRFWJKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Vázquez et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 74, 349-359 (2014-12-03)
The mercurial forms [inorganic divalent mercury, Hg(II) and methylmercury, CH3Hg] produce neurological and immune effects as well as hematological and renal alterations. The main route of exposure is through the diet. Consequently, the gastrointestinal mucosa is exposed to these mercurial
Yu Zhu et al.
Molecular pharmaceutics, 12(2), 332-341 (2014-08-26)
Polycations are explored as carriers to deliver therapeutic nucleic acids. Polycations are conventionally pharmacological inert with the sole function of delivering therapeutic cargo. This study reports synthesis of a self-immolative polycation (DSS-BEN) based on a polyamine analogue drug N(1),N(11)-bisethylnorspermine (BENSpm).
Swati Naphade et al.
Stem cells (Dayton, Ohio), 33(1), 301-309 (2014-09-05)
Despite controversies on the potential of hematopoietic stem cells (HSCs) to promote tissue repair, we previously showed that HSC transplantation could correct cystinosis, a multisystemic lysosomal storage disease, caused by a defective lysosomal membrane cystine transporter, cystinosin (CTNS gene). Addressing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service