Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

S2130

Sigma-Aldrich

5-Sulfosalicylic acid dihydrate

ReagentPlus®, ≥99%

Synonym(s):

2-Hydroxy-5-sulfobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO3SC6H3-2-(OH)CO2H·2H2O
CAS Number:
Molecular Weight:
254.21
Beilstein:
650741
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Quality Level

product line

ReagentPlus®

Assay

≥99%

mp

105-110 °C (lit.)

solubility

water: soluble 100 mg/mL, clear, colorless

functional group

carboxylic acid
sulfonic acid

SMILES string

[H]O[H].[H]O[H].OC(=O)c1cc(ccc1O)S(O)(=O)=O

InChI

1S/C7H6O6S.2H2O/c8-6-2-1-4(14(11,12)13)3-5(6)7(9)10;;/h1-3,8H,(H,9,10)(H,11,12,13);2*1H2

InChI key

BHDKTFQBRFWJKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

5-Sulfosalicylic acid dihydrate is a poly functional metal chelating ligand that may be used to form metal coordination complexes.

Application


  • Efficient transfer hydrogenation of biomass-derived furfural to furfuryl alcohol over mesoporous Zr-containing hybrids: This research uses 5-sulfosalicylic acid as a ligand to enhance catalytic efficiency, demonstrating potential applications in biofuel production (J Yang et al., 2022).

  • Synthesis of polyaniline/5-sulfosalicylic acid composite and its electrochemical properties: Investigates the synthesis of a composite material that exhibits promising electrochemical properties, useful for developing advanced electronic materials (RM Bandeira et al., 2020).

  • Controllable 5-sulfosalicylic acid assisted solvothermal synthesis of monodispersed superparamagnetic Fe3O4 nanoclusters: Highlights the role of 5-sulfosalicylic acid in controlling the size and dispersion of magnetic nanoparticles, crucial for various technological applications (W Wang et al., 2017).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, structure, and physical properties of calcium and strontium complexes with the ligand sulfosalicylic acid.
Wu G, et al.
Zeitschrift fur Anorganische und Allgemeine Chemie, 636(3?4), 652-656 (2010)
Syntheses, structures, and characterizations of four new lead (II) 5-sulfosalicylate complexes with both chelating and bridging neutral ligands.
Fan SR, & Zhu LG.
Inorganic Chemistry, 46(16), 6785-6793 (2007)
Kamil Strzelak et al.
Biosensors & bioelectronics, 127, 31-37 (2018-12-28)
The fully-mechanized Multicommutated Flow Analysis (MCFA) system for determination of transferrin in human serum has been developed. The analytical methodology concerns immunoprecipitation measurements with the use of LED-based nephelometric flow-through detectors with polymeric light guides. To improve the mechanization degree
Butyltin (IV) 5-sulfosalicylates: structural characterization and their cytostatic activity
Pruchnik H, et al
Polyhedron, 49(1), 223-233 (2013)
Madalina Andreea Badea et al.
SLAS discovery : advancing life sciences R & D, 24(5), 563-578 (2019-03-22)
This study aimed to develop and compare single and multiple 3D models such as multicellular tumor spheroids and to investigate the influence of Matrigel on their morphological and functional behavior. MDA-MB-231 3D models were generated in the presence and absence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service