Skip to Content
MilliporeSigma
All Photos(1)

Documents

28-5435

Sigma-Aldrich

Styrene

SAJ first grade, ≥99.0%

Synonym(s):

Phenylethylene, Vinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CH2
CAS Number:
Molecular Weight:
104.15
Beilstein:
1071236
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

SAJ first grade

vapor density

3.6 (vs air)

vapor pressure

12.4 mmHg ( 37.7 °C)
4.3 mmHg ( 15 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

914 °F

expl. lim.

6.1 %

availability

available only in Japan

refractive index

n20/D 1.546 (lit.)

bp

145-146 °C (lit.)

mp

−31 °C (lit.)

density

0.906 g/mL at 25 °C

SMILES string

C=Cc1ccccc1

InChI

1S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7H,1H2

InChI key

PPBRXRYQALVLMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

hearing organs, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shunsuke Tanaka et al.
ACS applied materials & interfaces, 10(1), 1039-1049 (2017-12-01)
Herein, we report the soft-templated preparation of mesoporous iron oxide using an asymmetric poly(styrene-b-acrylic acid-b-ethylene glycol) (PS-b-PAA-b-PEG) triblock copolymer. This polymer forms a micelle consisting of a PS core, a PAA shell, and a PEG corona in aqueous solutions, which
Niall D O'Leary et al.
FEMS microbiology reviews, 26(4), 403-417 (2002-11-05)
The last few decades have seen a steady increase in the global production and utilisation of the alkenylbenzene, styrene. The compound is of major importance in the petrochemical and polymer-processing industries, which can contribute to the pollution of natural resources
Bum Gun Kwon et al.
Environmental pollution (Barking, Essex : 1987), 188, 45-49 (2014-02-21)
Beach sand and seawater taken from the coastlines of the North-East Pacific Ocean and Hawaii State were investigated to determine the causes of global chemical contamination from polystyrene (PS). All samples were found to contain styrene monomer (SM), styrene dimers (SD)
Pavel Vodicka et al.
Mutation research, 511(3), 239-254 (2002-06-29)
Styrene is an important industrial chemical that has shown genotoxicity in many toxicology assays. This is believed to be related to the DNA-binding properties of styrene-7,8-oxide (SO), a major metabolite of styrene. In this review, we have summarized knowledge on
James J Collins et al.
Epidemiology (Cambridge, Mass.), 24(2), 195-203 (2013-01-25)
Epidemiologic studies have reported increased risk of lymphohematopoietic cancers, lung cancer, and pancreatic cancer after exposure to styrene, although findings across studies are not consistent. We update a large study of reinforced plastic industry workers with relatively high exposures to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service