MilliporeSigma
  • Spectrum of styrene-induced DNA adducts: the relationship to other biomarkers and prospects in human biomonitoring.
Spectrum of styrene-induced DNA adducts: the relationship to other biomarkers and prospects in human biomonitoring.
Mutation research (2002-06-29)
Pavel Vodicka, Mikko Koskinen, Michael Arand, Franz Oesch, Kari Hemminki
ABSTRACT
Styrene is an important industrial chemical that has shown genotoxicity in many toxicology assays. This is believed to be related to the DNA-binding properties of styrene-7,8-oxide (SO), a major metabolite of styrene. In this review, we have summarized knowledge on various aspects of styrene genotoxicity, especially in order to understand the formation and removal of primary DNA lesions, and the usefulness of biomarkers for risk assessment. Biological significances of specific DNA adducts and their role in the cascade of genotoxic events are discussed. Links between markers of external and internal exposure are evaluated, as well as metabolic aspects leading to the formation of DNA adducts and influencing biomarkers of biological effect. Finally, we suggest a design of a population study, which may contribute to our understanding genotoxic events in the exposure either to single xenobiotic or complex mixture.
Sigma-Aldrich
Styrene, SAJ first grade, ≥99.0%
Supelco
Styrene, analytical standard
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Supelco
Styrene, analytical standard