Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-2700

Sigma-Aldrich

Sodium fluoride

JIS special grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaF
CAS Number:
Molecular Weight:
41.99
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS special grade

vapor pressure

1.4 mmHg ( 0 °C)

Assay

≥99.0%

form

solid

availability

available only in Japan

mp

993 °C (lit.)

storage temp.

15-25°C

SMILES string

[F-].[Na+]

InChI

1S/FH.Na/h1H;/q;+1/p-1

InChI key

PUZPDOWCWNUUKD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonja Verena Bergner et al.
Plant physiology, 168(2), 615-634 (2015-04-11)
In plants and algae, the serine/threonine kinase STN7/STT7, orthologous protein kinases in Chlamydomonas reinhardtii and Arabidopsis (Arabidopsis thaliana), respectively, is an important regulator in acclimation to changing light environments. In this work, we assessed STT7-dependent protein phosphorylation under high light
Naoomi Tominaga et al.
Nature communications, 6, 6716-6716 (2015-04-02)
Brain metastasis is an important cause of mortality in breast cancer patients. A key event during brain metastasis is the migration of cancer cells through blood-brain barrier (BBB). However, the molecular mechanism behind the passage through this natural barrier remains
Ryan D Espy et al.
Analytical chemistry, 86(15), 7712-7718 (2014-06-28)
Determination of eight drugs of abuse in blood has been performed using paper spray or extraction spray mass spectrometry in under 2 min with minimal sample preparation. A method has been optimized for quantification of amphetamine, methamphetamine, 3,4-methylenedioxyamphetamine (MDA), 3,4-methylenedioxy-N-methylamphetamine
Jelke J Fros et al.
The Journal of general virology, 96(Pt 3), 580-589 (2014-11-15)
The unfolded protein response (UPR) is a cellular defence mechanism against high concentrations of misfolded protein in the endoplasmic reticulum (ER). In the presence of misfolded proteins, ER-transmembrane proteins PERK and IRE1α become activated. PERK phosphorylates eIF2α leading to a
Pierre-Olivier Strale et al.
The Journal of cell biology, 210(2), 333-346 (2015-07-22)
Oligomerization of cadherins could provide the stability to ensure tissue cohesion. Cadherins mediate cell-cell adhesion by forming trans-interactions. They form cis-interactions whose role could be essential to stabilize intercellular junctions by shifting cadherin clusters from a fluid to an ordered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service