Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.72576

SAFC

Sodium fluoride

extra pure, for table water

Synonym(s):

Sodium fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
FNa
CAS Number:
Molecular Weight:
41.99
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-667-8

vapor pressure

1 hPa ( 1077 °C)

Quality Level

form

crystals

pH

10.2 (20 °C, 40 g/L in H2O)

mp

996 °C

solubility

soluble 42 g/L

density

2.8 g/cm3 at 20 °C

application(s)

flavors and fragrances
pharma/biopharma processes

storage temp.

2-30°C

InChI

1S/FH.Na/h1H;/q;+1/p-1

InChI key

PUZPDOWCWNUUKD-UHFFFAOYSA-M

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Sodium fluoride with a low bromide level can be used to remineralize desalinated sea water to produce table water.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service