Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-5170

Sigma-Aldrich

Potassium permanganate solution

0.02 M

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KMnO4
CAS Number:
Molecular Weight:
158.03
MDL number:
UNSPSC Code:
12352312
PubChem Substance ID:
form:
liquid

form

liquid

reaction suitability

reagent type: oxidant

availability

available only in Japan

concentration

0.02 M
1/10 N

SMILES string

[K][Mn](=O)(=O)(=O)=O

InChI

1S/K.Mn.4O/q+1;;;;;-1

InChI key

VZJVWSHVAAUDKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Darryl B Jones et al.
Water research, 46(17), 5491-5498 (2012-08-15)
In this study, the impacts of three preoxidation strategies [i.e., using potassium permanganate (KMnO(4)), chlorine dioxide (ClO(2)), or hydrogen peroxide (H(2)O(2))] before preformed monochloramine (NH(2)Cl) addition on the formation and speciation of iodinated trihalomethanes (I-THMs) were evaluated at the Br(-)/I(-)
Jenny Li-Ying Huang et al.
Cancer research, 75(16), 3384-3397 (2015-06-27)
GNAQ and GNA11 are heterotrimeric G protein alpha subunits, which are mutated in a mutually exclusive pattern in most cases of uveal melanoma, one of the most aggressive cancers. Here we introduce the first transgenic mouse model of uveal melanoma
F C Holstege et al.
Methods (San Diego, Calif.), 12(3), 203-211 (1997-07-01)
Open complex formation precedes initiation of transcription by RNA polymerases. In the analysis of transcription initiation from eukaryotic class II promoters, we have used promoter DNA structures that represent intermediates in open complex formation. We describe the preparation and isolation
Eun-Joo Shin et al.
Free radical biology & medicine, 84, 355-372 (2015-04-07)
To determine the role of ceruloplasmin (Cp) in epileptic seizures, we used a kainate (KA) seizure animal model and examined hippocampal samples from epileptic patients. Treatment with KA resulted in a time-dependent decrease in Cp protein expression in the hippocampus
Zhanchao Meng et al.
Nature communications, 6, 6096-6096 (2015-02-05)
Indolosesquiterpenoids are a growing class of natural products that exhibit a wide range of biological activities. Here, we report the total syntheses of xiamycin A and oridamycins A and B, indolosesquiterpenoids isolated from Streptomyces. Two parallel strategies were exploited to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service