19-3480
Methyl 4-hydroxybenzoate
SAJ first grade, ≥98.0%
Synonym(s):
p-Hydroxybenzoic acid methyl ester, Methyl paraben, NIPAGIN
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HOC6H4CO2CH3
CAS Number:
Molecular Weight:
152.15
Beilstein:
509801
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
grade:
SAJ first grade
Recommended Products
grade
SAJ first grade
Assay
≥98.0%
form
solid
availability
available only in Japan
dilution
(for analytical testing)
mp
125-128 °C (lit.)
SMILES string
COC(=O)c1ccc(O)cc1
InChI
1S/C8H8O3/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5,9H,1H3
InChI key
LXCFILQKKLGQFO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
334.4 °F
Flash Point(C)
168 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Martyna Pajewska-Szmyt et al.
Materials (Basel, Switzerland), 13(19) (2020-10-03)
Magnetic molecularly imprinted polymers (MMIPs) are an invaluable asset in the development of many methods in analytical chemistry, particularly sample preparation. Novel adsorbents based on MMIPs are characterized by high selectivity towards a specific analyte due to the presence of
Ismail I Hewala et al.
Talanta, 88, 623-630 (2012-01-24)
Simple methods for HPTLC peak purity assessment and identification of the HPTLC peaks were presented. The spectrodensitograms - selected at different time intervals across the elution time of the HPTLC peak - were extracted and digital algorithms for manipulating the
Gabriela Oliveira et al.
International journal of pharmaceutics, 435(1), 38-49 (2012-05-29)
Simple topical formulations which include volatile components, such as gels or sprays, are appealing from a cosmetic perspective. However, complex formulation effects may result from the use of volatile excipients in topical formulations, particularly when applied at clinically relevant doses
Kyung-Jin Min et al.
Aging cell, 7(2), 199-206 (2008-01-29)
Reduced insulin/insulin-like growth factor (IGF) signaling may be a natural way for the reduction of dietary nutrients to extend lifespan. While evidence challenging this hypothesis is accumulating with Caenorhabditis elegans, for Drosophila melanogaster it is still thought that insulin/IGF and
K Kannathasan et al.
Acta tropica, 120(1-2), 115-118 (2011-07-19)
The vector-borne diseases caused by mosquitoes are one of the major health problems in many countries especially in tropical and sub-tropical countries. The resistance of mosquitoes to synthetic chemicals and environmental toxicity created by the chemicals raised the demand for
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service