Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18-1200

Sigma-Aldrich

Ligroin

SAJ first grade, ≥90.0%

Synonym(s):

Mineral spirits, Petroleum spirits

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12000000

grade

SAJ first grade

vapor density

>1 (vs air)

Assay

≥90.0%

form

liquid

availability

available only in Japan

refractive index

n20/D 1.376 (lit.)

bp

60-80 °C (lit.)

density

0.656 g/mL at 25 °C (lit.)

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Muta. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-56.2 °F - closed cup

Flash Point(C)

-49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catherine F Poh et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 12(22), 6716-6722 (2006-11-24)
Genetically altered cells could become widespread across the epithelium of patients with oral cancer, often in clinically and histologically normal tissue, and contribute to recurrent disease. Molecular approaches have begun to yield information on cancer/risk fields; tissue optics could further
I P Dick et al.
Human & experimental toxicology, 16(11), 645-651 (1998-01-14)
1. The absorption and distribution of lindane through skin was examined using human volunteers. Two different preparations were employed, one with acetone as the vehicle and the other, a commercial product, consisting primarily of white spirit as the solvent base.
The use of a surgical glove in the prevention of tourniquet-associated chemical burns.
P J Tomlinson et al.
Annals of the Royal College of Surgeons of England, 90(3), 255-255 (2008-04-26)
I P Dick et al.
Human & experimental toxicology, 16(11), 652-657 (1998-01-14)
1. The absorption of lindane through human skin was assessed in vitro using static diffusion cells with 50% aqueous ethanol as the receptor fluid. Four formulations of lindane were applied, three of which were commercial preparations. The dermal distribution of
A Steensgaard et al.
Neurotoxicology, 17(3-4), 785-792 (1996-10-01)
Several epidemiological studies of workers occupationally exposed to white spirit show that neuropsychiatric disorders are a frequent cause of early disability pension in this population compared with non-exposed controls. In the rat, we have demonstrated that exposure to different kinds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service