Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
product line
TraceCERT®
grade
certified reference material, TraceCERT®
composition
nitrate, 1 g/L , phosphate, 1 g/L , sulfate, 1 g/L
concentration
in water
technique(s)
ion chromatography: suitable
application(s)
environmental
food and beverages
format
multi-component solution
Analysis Note
Other Notes
It contains Nitrate (NO3-), Phosphate (PO43-)and Sulfate (SO42-) at nominal concentrations of 1000 mg/L each
Visit the IC Portal to learn more
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable |
| application(s) environmental | application(s) environmental | application(s) environmental | application(s) environmental |
| format multi-component solution | format multi-component solution | format multi-component solution | format multi-component solution |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® |
| concentration in water | concentration in water | concentration in 0.1% nitric acid | concentration in water |
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



