All Photos(1)
About This Item
Linear Formula:
AlCuZn
CAS Number:
MDL number:
UNSPSC Code:
12352402
PubChem Substance ID:
Recommended Products
grade
SAJ special grade
form
powder
composition
aluminum, ~45%
copper, ~50%
zinc, ~5%
availability
available only in Japan
mp
550 °C (lit.)
SMILES string
[Al].[Cu].[Zn]
InChI
1S/Al.Cu.Zn
InChI key
MUBKMWFYVHYZAI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service