Skip to Content
MilliporeSigma
All Photos(1)

Documents

ABE1338

Sigma-Aldrich

Anti-METTL14 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

N6-adenosine-methyltransferase subunit METTL14, m6A methyltransferase METTL14, Methyltransferase-like protein 14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

zebrafish (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... METTL14(57721)

General description

N6-adenosine-methyltransferase subunit METTL14 (EC 2.1.1.62; UniProt Q9HCE5; also known as m6A methyltransferase METTL14, Methyltransferase-like protein 14) is encoded by the METTL14 (also known as KIAA1627) gene (Gene ID 57721) in human. N(6)-Methyladenosine (m(6)A) represents the most prevalent internal modification in messenger and long noncoding RNAs. This reversible modification is installed by the m(6)A methylation "writers" and remomved by demethylases that serve as "erasers. In addition, methyl-specific binding proteins, primarily of the YTH-domain family, function as "readers" and mediate the effect by binding to modified transcripts. METTL14 is a component of the nuclear methyltransferase complex (writers) composed of METTL3, METTL14, and WTAP. At least 2 demethylases (erasers), ALKBH5 and FTO, that can catalyze the reverse reaction. The roles of m(6)A in cell fates regulation and embryonic development highlight the existence of another layer of epigenetic regulation at the RNA level.

Specificity

The immunogen sequence is present in human METTL14 (UniProt Q9HCE5) and both of murine METTL14 spliced isoforms (Uniprot Q3UIK4).

Immunogen

KLH-conjugated linear peptide corresponding to an internal sequence from the C-terminal half of human METTL14.

Application

This Anti-METTL14 Antibody is validated for use in Western Blotting for the detection of METTL14.

Quality

Evaluated by Western Blotting in Raji nuclear extract.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected METTL14 in 10 µg of Raji nuclear extract.

Target description

~56 kDa observed. 52.15 kDa (human), 52.12/40.72 kDa (mouse isoform 1/2) calculated. Uncharacterized band(s) may appear in some lysates.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loss of METTL3 attenuates blastic plasmacytoid dendritic cell neoplasm response to PRMT5 inhibition via IFN signaling.
Rethnam, et al.
Blood advances, 6, 5330-5344 (2023)
Fanpeng Zhao et al.
Molecular neurodegeneration, 16(1), 70-70 (2021-10-02)
N6-methyladenosine (m6A) modification of RNA influences fundamental aspects of RNA metabolism and m6A dysregulation is implicated in various human diseases. In this study, we explored the potential role of RNA m6A modification in the pathogenesis of Alzheimer disease (AD). We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service