Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-6450

Sigma-Aldrich

Cresol mixture of isomers

CP

Synonym(s):

Methylphenol tricresol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

CP

form

liquid

availability

available only in Japan

SMILES string

Cc1ccc(O)cc1.Cc2cccc(O)c2.Cc3ccccc3O

InChI

1S/3C7H8O/c1-6-2-4-7(8)5-3-6;1-6-3-2-4-7(8)5-6;1-6-4-2-3-5-7(6)8/h3*2-5,8H,1H3

InChI key

QTWJRLJHJPIABL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Himansu S Biswal et al.
The Journal of chemical physics, 129(18), 184311-184311 (2008-12-03)
Experimental evidence is presented for the O-H-S hydrogen bonding in the complexes of simple model compounds of methionine (dimethyl sulphide) and tyrosine (phenol, p-cresol, and 2-naphthol). The complexes were formed in the supersonic jet and were detected using resonantly enhanced
Alfredo Gallego et al.
Environmental toxicology, 23(6), 664-671 (2008-02-23)
We studied the degradation of mixtures of o-cresol, m-cresol, and p-cresol, by Pseudomonas putida isolated from natural sources, and the application of this degradation to the depuration and detoxification of synthetic and industrial wastewater. Biodegradation assays were performed in batch
Koji Okamoto et al.
BMJ case reports, 2011, doi:10-doi:10 (2011-01-01)
The authors report a case of a 43-year-old woman who presented with second degree chemical burns to 9% of the total body surface area due to cutaneous contact with cresol. This was associated with acute oliguric kidney injury requiring haemodialysis.
Paweena Susantitaphong et al.
Blood purification, 28(2), 93-101 (2009-05-15)
On-line hemodiafiltration (OL-HDF) with pre- as well as post-dilution reinfusion mode provides superior uremic toxin removal, especially middle uremic toxins, and can improve patient survival. Both standard pre- and post-dilution have limitations. Mid-dilution OL-HDF (Nephros OLpur) was invented to overcome
Kazuhide Ueno et al.
The journal of physical chemistry. B, 115(48), 13994-13999 (2011-07-07)
To support a new interpretation of the origin of the dynamic heterogeneity observed pervasively in fragile liquids as they approach their glass transition temperatures T(g), we demonstrate that the introduction of ~2 nm structural inhomogeneities into a homogeneous glass former

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service