MilliporeSigma
Acute tubular necrosis due to cutaneous contact with cresol.
BMJ case reports (2011-01-01)
Koji Okamoto, Eisei Noiri, Machiko Oka, Hidekazu Moriya, Takayasu Ohtake, Shuzo Kobayashi
ABSTRACT
The authors report a case of a 43-year-old woman who presented with second degree chemical burns to 9% of the total body surface area due to cutaneous contact with cresol. This was associated with acute oliguric kidney injury requiring haemodialysis. In contrast to previous reports of cresol ingestion, the patient did not have evidence of hepatic dysfunction, possibly due to a low cresol concentration in the portal vein and liver. Renal histopathology showed regional accentuated tubular necrosis and disruption of the tubular basement membrane. Renal toxicity was thought to be due to direct tubular toxicity and impaired renal blood flow.
Sigma-Aldrich
Cresol mixture of isomers, technical, crude, ~85% (sum of isomers, GC)
Sigma-Aldrich
Cresol mixture of isomers, CP