Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01-0870

Sigma-Aldrich

N-Acetyl-L-cysteine

SAJ special grade, 98.0-102.0%

Synonym(s):

LNAC, NAC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(NHCOCH3)CO2H
CAS Number:
Molecular Weight:
163.19
Beilstein:
1724426
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

grade

SAJ special grade

Assay

98.0-102.0%

form

solid

availability

available only in Japan

mp

106-108 °C (lit.)

application(s)

peptide synthesis

shipped in

wet ice

storage temp.

2-8°C

SMILES string

CC(=O)N[C@@H](CS)C(O)=O

InChI

1S/C5H9NO3S/c1-3(7)6-4(2-10)5(8)9/h4,10H,2H2,1H3,(H,6,7)(H,8,9)/t4-/m0/s1

InChI key

PWKSKIMOESPYIA-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Antioxidant and mucolytic agent. Increases cellular pools of free radical scavengers. Reported to prevent apoptosis in neuronal cells but induce apoptosis in smooth muscle cells. Inhibits HIV replication. May serve as a substrate for microsomal glutathione transferase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suzanne M Cloonan et al.
Journal of medicinal chemistry, 58(11), 4494-4505 (2015-05-12)
Ruthenium polypyridyl complexes show great promise as new photodynamic therapy (PDT) agents. However, a lack of detailed understanding of their mode of action in cells poses a challenge to their development. We have designed a new Ru(II) PDT candidate that
Eduesley Santana-Santos et al.
The Annals of thoracic surgery, 97(5), 1617-1623 (2014-03-25)
The renoprotective effect of N-acetylcystein in patients undergoing coronary artery bypass graft surgery is controversial. We assessed the renoprotective effect of the highest dose of N-acetylcystein sanctioned for clinical use in a prospective, double-blind, placebo-controlled study including 70 chronic kidney
Michael Berk et al.
Trends in pharmacological sciences, 34(3), 167-177 (2013-02-02)
N-Acetylcysteine (NAC) targets a diverse array of factors germane to the pathophysiology of multiple neuropsychiatric disorders including glutamatergic transmission, the antioxidant glutathione, neurotrophins, apoptosis, mitochondrial function, and inflammatory pathways. This review summarises the areas where the mechanisms of action of
Anna M Sadowska
Therapeutic advances in respiratory disease, 6(3), 127-135 (2012-03-01)
To develop an efficient therapy for chronic obstructive pulmonary disease (COPD), N-acetylcysteine (NAC) has been tested as a medication that can suppress various pathogenic processes in this disease. NAC is a thiol compound, which provides sulfhydryl groups. NAC can act
Yuval Samuni et al.
Biochimica et biophysica acta, 1830(8), 4117-4129 (2013-04-27)
N-acetylcysteine (NAC) has been in clinical practice for several decades. It has been used as a mucolytic agent and for the treatment of numerous disorders including paracetamol intoxication, doxorubicin cardiotoxicity, ischemia-reperfusion cardiac injury, acute respiratory distress syndrome, bronchitis, chemotherapy-induced toxicity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service