Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

106425

Millipore

N-Acetyl-L-cysteine

Mucolytic agent and thiol-based antioxidant that has a protective effect against oxygen free radicals in early septic shock.

Synonym(s):

N-Acetyl-L-cysteine, LNAC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO3S
CAS Number:
Molecular Weight:
163.19
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.25

description

Merck USA index - 14, 88

Quality Level

Assay

≥98% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 50 mg/mL

anion traces

chloride (Cl-): ≤0.04%
sulfate (SO42-): ≤0.03%

cation traces

Fe: ≤20 ppm

shipped in

ambient

storage temp.

15-25°C

SMILES string

SCC(NC(=O)C)C(=O)O

InChI

1S/C5H9NO3S/c1-3(7)6-4(2-10)5(8)9/h4,10H,2H2,1H3,(H,6,7)(H,8,9)

InChI key

PWKSKIMOESPYIA-UHFFFAOYSA-N

General description

A mucolytic agent and thio-based antioxidant. Increases the cellular pool of free-radical scavengers. Inhibits HIV replication. Prevents apoptotic death of neuronal cells. Antidote for acetaminophen poisoning. May serve as a substrate for microsomal glutathione transferase. Induces apoptosis in human and rat smooth muscle cells.
Mucolytic agent and thiol-based antioxidant that has a protective effect against oxygen free radicals in early septic shock. Increases cellular pools of free-radical scavengers. Inhibitor of HIV replication. May serve as a substrate for microsomal glutathione transferase. Induces apoptosis in human and rat smooth muscle cells.

Application


  • Chlorpyrifos-induced suppression of the antioxidative defense system leads to cytotoxicity and genotoxicity in macrophages.: The study highlights the protective role of N-acetyl-L-cysteine against chlorpyrifos-induced oxidative damage in macrophages, emphasizing its importance in mitigating pesticide toxicity (Lu et al., 2024).

  • N-Acetyl-L-Cysteine Attenuates Titanium Dioxide Nanoparticle (TiO2 NP)-Induced Autophagy in Male Germ Cells.: This research explores how N-acetyl-L-cysteine can mitigate the autophagic effects induced by titanium dioxide nanoparticles in male germ cells, suggesting its use in reproductive health (Shin et al., 2024).


Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Aqueous stock solutions are stable for up to 1 month at -20°C.

Other Notes

Ortolani, O., et al. 2000. Am. J. Respir. Crit. Care Med. 161, 1907.
Tsai, J.C., et al. 1996. J. Biol. Chem. 271, 3667.
Ferrari, G., et al. 1995. J. Neurosci.15, 2857.
Weinander, R., et al. 1994. J. Biol. Chem.269, 71.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service