Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11934317001

Roche

RNA/DNA Stabilization Reagent for Blood/Bone Marrow

Synonym(s):

nucleic acid stabalization

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500

packaging

pkg of 500 mL

Quality Level

manufacturer/tradename

Roche

General description

For simultaneous cell lysis and stabilization of nucleic acids in blood or bone marrow samples.

Application

The RNA/DNA Stabilization Reagent for Blood/Bone Marrow is intended for general laboratory use and aids in the collection of samples for nucleic-acid isolation. It is able to lyse cells and simultaneously inactivate nucleases when added to blood or bone-marrow samples. In addition to providing safe transport from the collection site to the laboratory, the reagent also serves as an effective long-term preserving agent, protecting nucleic acids at –15 to –25°C until future analysis.
The easy-to-use reagent increases the accuracy of nucleic-acid detection by ensuring complete and immediate inactivation of nucleases. At the time of collection, one volume of blood sample or bone-marrow aspirate is added to 10 volumes of reagent, thus minimal reagent calculations are needed.

Quality

The reagent is function-tested in the following model system: the detection of the equivalent of five melanoma cells in a volume of 5 mL of normal human blood.
The recovery of tyrosinase mRNA is detected by RT-PCR after purification of 5 pg of MelJu cell mRNA, added to a lysate prepared of 5 mL of fresh normal human blood, and stabilized with the RNA/DNA Stabilization Reagent for Blood/Bone Marrow. The mRNA is purified from the stabilized lysate with the mRNA Isolation Kit for Blood/ Bone Marrow.
Note: 5 pg of MelJu cell mRNA is equivalent to about five melanoma cells.

Preparation Note

The RNA/DNA Stabilization Reagent for Blood/Bone Marrow contains a strong protein denaturant (e.g., guanidinium thiocyanate), detergents, and reducing agents to accomplish instantaneous cell lysis and nuclease inactivation. The nucleic acids are stabilized, and ready for transport back to the laboratory for isolation with the mRNA Isolation Kit for Blood/Bone Marrow.
Lysates can be immediately used, stored for one day at +2 to +8°C, or stored for up to 1 year at –15 to –25°C. Five hundred milliliters is sufficient to stabilize a total of 50 mL of whole blood or bone-marrow aspirate.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service