Skip to Content
MilliporeSigma
All Photos(1)

Documents

11699105001

Roche

PCR Buffer Without MgCl2, 10x concentrated

solution, pkg of 3 × 1 mL

Synonym(s):

buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105600

form

solution

Quality Level

packaging

pkg of 3 × 1 mL

manufacturer/tradename

Roche

storage temp.

−20°C

General description

The PCR buffer without MgCl2, in combination with the MgCl2 stock solution, is used for the individual adjustment of the Mg2+ concentration in the PCR reaction. In most applications a concentration of 1.5mM MgCl2 will yield satisfactory results with a dNTP concentration of 200μM each. In many cases, however, it is required to titrate the optimal Mg2+ concentration to increase the specificity and yield of the PCR reaction.The buffer is ten-fold concentrated and should be used in combination with the MgCl2 stock solution.

Application

The PCR buffer without MgCl2 has been used for the amplification of specific DNA fragments by the polymerase chain reaction (PCR).

Physical form

Solution, 100 mM Tris-HCl, 500 mM KCl (pH 8.3 at 20 °C)

Other Notes

For general laboratory use.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Omid Yeganeh et al.
Iranian Red Crescent medical journal, 15(4), 340-344 (2013-10-02)
Chlamydia trachomatis and Mycoplasma genitalium infections are the most prevalent sexually transmitted bacterial infections in the world that cause urogenital infections in both men and women. It appears that infertility is a complication of these infections. This study was designed
Rao L Divi et al.
Toxicological sciences : an official journal of the Society of Toxicology, 118(1), 191-201 (2010-08-13)
Mitochondrial compromise has been documented in infants born to women infected with the human immunodeficiency virus (HIV-1) who received nucleoside reverse transcriptase inhibitor (NRTI) therapy during pregnancy. To model these human exposures, we examined mitochondrial integrity at birth and 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service