Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11004638001

Roche

Chymostatin

Synonym(s):

Chymostatin, N-(Nα-Carbonyl-Cpd-X-Phe-al)-Phe (Cpd = capreomycidine) (capreomycidine = [S,S]-α-(2-Iminohexahydro-4-pyrimidyl)glycine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

form

powder

Quality Level

mol wt

Mr = 607.71

packaging

pkg of 10 mg

manufacturer/tradename

Roche

mp

205 °C

solubility

acetic acid: soluble 20 mg/mL

storage temp.

2-8°C

SMILES string

OC(C(NC(NC(C1NC(NCC1)=N)C([F,Cl,Br,I]C)=O)=O)CC2=CC=CC=C2)=O

InChI key

MRXDGVXSWIXTQL-HYHFHBMOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Chymostatin is a mixture of three components, A, B, and C. The component A being N-[((S)-1-carboxy-2-phenylethyl)-carbamoyl]-α-[2-iminohexahydro-4(S)-pyrimidyl]-L-glycyl-L-leucyl-phenylalaninal. The other two components B and C differ in that the L-leucyl residue is substituted by L-valine and L-isoleucine, respectively.

Application

Chymostatin is a specific inhibitor of α-, β-, γ-, and δ-chymotrypsin.

Biochem/physiol Actions

Chymostatin is a strong inhibitor of many proteases, including chymotrypsin, papain, chymotrypsin-like serine proteinases, chymases, and lysosomal cysteine proteinases such as cathepsins A,B,C, B, H, and L. It weakly inhibits human leucocyte elastase. It is effective at a final concentration of 100 to 200 μg/ml (10 to 100 μM). Chymostatin is often included in protease inhibitor cocktails used with plant extracts.

Quality

Performance tested.

Formula variant

C31H41N7O6

Preparation Note

Working concentration: 6 to 60 μg/ml (10 to 100 μM)
1 U chymotrypsin is inhibited to 49% of the original activity by 1.8 μg of chymostatin.
Thin-layer chromatography: butanol/methanol/H2O = 4 / 1 / 2
Working solution: Soluble in glacial acetic acid or DMSO to 20 mg/ml. Sparingly soluble in water, methanol, or ethanol. Insoluble in ethyl acetate, petroleum and ethyl ethers, hexane, or chloroform (CHCl3).
It is recommended to dissolve the inhibitor in 1% acetic acid in higher concentration and to adjust the concentration wanted with phosphate buffer, 0.05 M, pH 7.0, which is common for chymotrypsin assay.
CAUTION: DMSO (Dimethyl sulfoxide) will permeate the skin, carrying solubilized protease inhibitors. Always wear appropriate protection for eyes, skin, etc.
Storage conditions (working solution): -15 to -25 °C
Dilute solutions should be stored frozen in aliquots at -15 to -25 °C and are stable for approximately one month. Avoid repeated freezing. Growth of microorganisms should be avoided as proteases from microbial origin may hydrolyze the peptides.
Solubility testing in glacial acetic acid at 10 mg/ml yields a clear solution, which is usually colorless, but can be yellow in appearance. It is reportedly also soluble in DMSO; only slightly soluble in water and short-chain alcohols; insoluble in ethyl acetate, butyl acetate, ether, hexane, and petroleum ether. Stock solutions (10 mM) can be prepared in DMSO and are stable for months at -20 °C. Stock solutions can also be made in 0.1 M HCl. Dilute solutions (10-100 μM) are only stable for several hours, due to oxidation of the terminal aldehyde.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The structure of chymostatin, a chymotrypsin inhibitor.
K Tatsuta et al.
The Journal of antibiotics, 26(11), 625-646 (1973-11-01)
Verena van der Heide et al.
Cell reports, 38(11), 110508-110508 (2022-03-06)
Concerns that infection with severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), the etiological agent of coronavirus disease 2019 (COVID-19), may cause new-onset diabetes persist in an evolving research landscape, and precise risk assessment is hampered by, at times, conflicting evidence.
Mirza Muhammad Fahd Qadir et al.
Nature communications, 11(1), 3265-3265 (2020-07-01)
The culture of live pancreatic tissue slices is a powerful tool for the interrogation of physiology and pathology in an in vitro setting that retains near-intact cytoarchitecture. However, current culture conditions for human pancreatic slices (HPSs) have only been tested
Sarah Bui et al.
Methods in molecular biology (Clifton, N.J.), 2557, 453-493 (2022-12-14)
In this chapter, we provide a detailed guide for the application of commonly used small molecules to study Golgi structure and function in vitro. Furthermore, we have curated a concise, validated list of endomembrane markers typically used in downstream assays

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service