MilliporeSigma
Limited extent and consequences of pancreatic SARS-CoV-2 infection.
Cell reports (2022-03-06)
Verena van der Heide, Sonia Jangra, Phillip Cohen, Raveen Rathnasinghe, Sadaf Aslam, Teresa Aydillo, Daniel Geanon, Diana Handler, Geoffrey Kelley, Brian Lee, Adeeb Rahman, Travis Dawson, Jingjing Qi, Darwin D'Souza, Seunghee Kim-Schulze, Julia K Panzer, Alejandro Caicedo, Irina Kusmartseva, Amanda L Posgai, Mark A Atkinson, Randy A Albrecht, Adolfo García-Sastre, Brad R Rosenberg, Michael Schotsaert, Dirk Homann
ABSTRACT
Concerns that infection with severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), the etiological agent of coronavirus disease 2019 (COVID-19), may cause new-onset diabetes persist in an evolving research landscape, and precise risk assessment is hampered by, at times, conflicting evidence. Here, leveraging comprehensive single-cell analyses of in vitro SARS-CoV-2-infected human pancreatic islets, we demonstrate that productive infection is strictly dependent on the SARS-CoV-2 entry receptor ACE2 and targets practically all pancreatic cell types. Importantly, the infection remains highly circumscribed and largely non-cytopathic and, despite a high viral burden in infected subsets, promotes only modest cellular perturbations and inflammatory responses. Similar experimental outcomes are also observed after islet infection with endemic coronaviruses. Thus, the limits of pancreatic SARS-CoV-2 infection, even under in vitro conditions of enhanced virus exposure, challenge the proposition that in vivo targeting of β cells by SARS-CoV-2 precipitates new-onset diabetes. Whether restricted pancreatic damage and immunological alterations accrued by COVID-19 increase cumulative diabetes risk, however, remains to be evaluated.
Product Description
Roche
Chymostatin
Sigma-Aldrich
Anti-Collagen Type I Antibody, clone 5D8-G9, clone 5D8-G9, Chemicon®, from mouse
Sigma-Aldrich
Aprotinin from bovine lung, BioUltra, 3-8 TIU/mg solid, ≥98% (SDS-PAGE)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Coronavirus Group Antigen Antibody, nucleoprotein of OC-43, clone 542-7D, clone 542-7D, Chemicon®, from mouse