Skip to Content
MilliporeSigma
All Photos(3)

Documents

ZR0L52120

Milli-Q

Milli-Q® HR High-Flow RO Water Purification System

Central water purification system for up to 2400 L/day Type 3 water. For high chlorine feed water.

Synonym(s):

Milli-Q® HR 7120 High-Flow RO Water Purification System (HC)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104204
NACRES:
JB.11

AC/DC input

100 V / 230 V, 50-60 Hz

Quality Level

product line

Milli-Q® HR

feature

Ionic rejection by RO: 95 - 99%
Organic rejection by RO: >=99% for MW >= 200

packaging

pkg of 1 unit

parameter

120 L/hr make-up flow rate
2400 L/day max. usage
94 kg operating weight (208 lb)

system depth × H × W

54.2 cm (21.3 in.) × 124 cm (48.8 in.) × 54.3 cm (21.4 in.)

input

feed water nature potable tap water

output

product water quality: type 3 water (RO water)

General description

The Milli-Q® HR 7120 system is a high-throughput standard water purification solution that can reliably meet the diverse needs requiring reverse osmosis (RO, Type 3) pure water – for a single laboratory or an entire research facility. These systems can produce up to 2400 L/day of laboratory-grade water from potable feed water. This system is suitable for feed water that has a high chlorine content.
Milli-Q® HR 7000 series systems can be integrated into a centralized system, providing total control of all parameters within the system itself as well as within the external pure water distribution loop.
These central systems are embedded with our unique MyMilli-Q Remote Care online monitoring and service capability, which operates as an extension of Milli-Q® Services. This digital service lets you securely and remotely manage your system, supporting the lab′s optimal productivity.

Application

The RO/Type 3 water produced by Milli-Q® HR 7000 series systems is ideal for all your laboratory pure water needs such as glassware rinsing, heating or sonicating baths, and basic reagent preparation.
It can feed instruments, including dishwashers, autoclaves, humidity and environmental chambers, and ice machines.
It can also feed Milli-Q® ultrapure water purification systems.

Features and Benefits

  • Enhances sustainability and reduces running costs, making your facility more productive, environmentally friendly, and cost-effective.
  • Superior ergonomic design for quick and easy maintenance.
  • Progard® Pretreatment cartridges remove particles, free chlorine and colloids.
  • An interactive touchscreen with helpful wizards for ease of operation and maintenance.
  • Modern data management capabilities for greater accessibility and data traceability.
  • MyMilli-Q Remote Care monitoring and service capability.
  • Equipped with patented E.R.A.® (Evolutive Reject Adjustment) technology to save you water, time and money. It automatically assesses feed water quality to produce a constant flow rate and optimize water recovery (between 45 and 75%).
  • Compatible with SDS 500 storage, protection, and distribution system, for high storage and distribution flow possibilities.

Components

  • The indicated price currently corresponds to the above catalog number only; it is subject to change at any time without notice.
  • This catalog number does not include consumables, accessories, and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time.
  • Because our customers each have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you, do not hesitate to contact us!

Legal Information

E.R.A. is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
MyMilli-Q is a trademark of Merck KGaA, Darmstadt, Germany
PROGARD is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Experience efficient Milli-Q® HR 7000 Central RO Water Purification Systems. A more sustainable high-flow pure water system with versatile configurations and modern control. Advanced RO technology saves water. Online monitoring gives peace of mind. Request information now!

Experience efficient Milli-Q® HR 7000 Central RO Water Purification Systems. A more sustainable high-flow pure water system with versatile configurations and modern control. Advanced RO technology saves water. Online monitoring gives peace of mind. Request information now!

Experience efficient Milli-Q® HR 7000 Central RO Water Purification Systems. A more sustainable high-flow pure water system with versatile configurations and modern control. Advanced RO technology saves water. Online monitoring gives peace of mind. Request information now!

Experience efficient Milli-Q® HR 7000 Central RO Water Purification Systems. A more sustainable high-flow pure water system with versatile configurations and modern control. Advanced RO technology saves water. Online monitoring gives peace of mind. Request information now!

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service