Skip to Content
MilliporeSigma

CFIF01205

Kit: Milli-Q® Ion-Free Ultrapure Cartridge

For use with Milli-Q® Bowl systems

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
JA.13
UNSPSC Code:
41104212
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

packaging

pkg of 1 kit

manufacturer/tradename

Milli-Q®

General description

The Ultrapure Cartridge Kit is an all-purpose water purification consumable designed to remove trace organic and inorganic contaminants from RiOs, Elix® or distilled feed water.

Application

Removes ionized impurities from RiOs, Elix® or distilled feed water to produce high-quality ultrapure water.

Other Notes

Directions For Use:

  • Organism Retention: Microorganisms
  • Mode of Action: Filtering
  • Application: General laboratory analysis
  • Intended Use: Water purification
  • Instructions for Use: This item provides water filtered through 0.22 μm sterilizing-grade membranes. Refer to the system equipment user guide section "Using the System" (available on USB key in the system box).
  • Storage Statement: Store in a dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library