Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ZMQSFTS01

Milli-Q

Foot Pedal

Hands-free dispensing accessory for Milli-Q® Advantage A10 / Integral, Q-POD® Element, and Elix® Advantage systems

Synonym(s):

Foot Pedal for Milli-Q® and Elix® Water Systems

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104200
eCl@ss:
33050190
NACRES:
NA.21

feature

Remotely controls water delivery by a simple push of the foot.

packaging

pkg of 1 unit

manufacturer/tradename

Q-POD®

cable L

3 m

compatibility

for use with Elix® Advantage
for use with Milli-Q® Advantage A10
for use with Milli-Q® Integral
for use with Q-POD® Element

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

The foot pedal accessory remotely controls water delivery by a simple push of the foot, when connected to a Q-POD® ultrapure or E-POD® pure water dispenser. Once the required volume of water is reached, releasing the foot from the pedal instantly stops the flow.
When combined with Volumetric Dispense mode, one press on the pedal starts the production of purified water at the point of dispense. Dispensing automatically stops at the set volume.
Hands-free dispensing accessory for use with Milli-Q® Advantage A10, Milli-Q® Integral, Q-POD® Element, and Elix® Advantage systems.

Features and Benefits

  • Ideal for scientists who need their free hands when accessing purified water.
  • Volumetric dispense mode saves time when repeat dispensing the same volume.
  • Supports lab sanitary protocols as users do not need to touch the system to access water.
  • A 3 m cable provides flexibility to position the pedal according to your lab design constraints.

Legal Information

E-POD is a registered trademark of Merck KGaA, Darmstadt, Germany
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Q-POD is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service