Key Documents
ZIQ7ESP01
Milli-Q
Sanitary Valve Kit
Aseptic sampling valve, for use with Milli-Q® IQ/IX systems
About This Item
UNSPSC Code:
41104200
NACRES:
JA.13
Pricing and availability is not currently available.
feature
Asseptic Water Sampling
packaging
pkg of 1 unit
compatibility
for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015
General description
This accessory kit allows the sampling of recirculating water in aseptic conditions. Aseptic sampling valve to sample water from the Milli-Q® IQ 7003/05/10/15 and Milli-Q® IX 7003/05/10/15 recirculation loop.
Application
Aseptic sampling valve to sample water from the Milli-Q® IQ 7003/5/10/15 and Milli-Q® IX 7003/5/10/15 recirculation loop.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany