Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WMBWASHA1

Milli-Q

Wall Mounting Bracket

Installation accessory for use with Milli-Q® IQ and Milli-Q® IX systems

Synonym(s):

Wall Mounting Bracket for Washer Distribution Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104214
NACRES:
JA.13

feature

Save lab bench space

packaging

pkg of 1 unit

compatibility

for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015

Looking for similar products? Visit Product Comparison Guide

General description

The mounting bracket easily installs a Washer Distribution Kit high up on a wall, leaving benchtops free of clutter for optimized lab space and a more efficient working environment.
This bracket is compatible with:
  • Washer Distribution Kit 230 V (right)
  • Washer Distribution Kit 230 V (left)
  • Washer Distribution Kit 115 V (right)
  • Washer Distribution Kit 115 V (left)

Features and Benefits

  • Easy to install.
  • Saves lab bench space.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service