Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TMSTK

Millipore

mPAGE® TurboMix Stacking Solution 120ML

Premixed stacking gel solution for casting bis-tris polyacrylamide gels

Synonym(s):

Bis-Tris gel stacking solution, stacking gel solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319
NACRES:
NA.25

Quality Level

sterility

0.2 μm filtered

form

liquid

feed ratio

29:1

storage condition

protect from light

storage temp.

2-8°C

General description

mPAGE® TurboMix Stacking Solution is a 4% acrylamide buffer solution used to make bis-tris polyacrylamide gels. It is a ready-to-use solution for stacking gels. First, add APS and TEMED. After pouring the resolving gel, pour the stacking solution and insert your comb.

Application

Bis-Tris Stacking Gel Solution
Premixed stacking gel solution for casting bis-tris polyacrylamide gels

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service