Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SYMBSBH01

Millipore

Steritest® Symbio Slim Bottle Holder

With Fastening Screw, for use with Steritest® Symbio pump that streamlines the workflow and increases safety

Synonym(s):

Bottle mount

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
NACRES:
NB.51

material

316L stainless steel holder

Agency

complies with EP (2.6.1)
complies with JP (4.06)
complies with USP (71)

manufacturer/tradename

Steritest®

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio Pump

shipped in

ambient

General description

The Steritest® Symbio Slim Bottle Holder is an accessory for the Steritest® Symbio pumps. Steritest® Symbio Pumps are used for reliable, faster, and easy sterility testing in a laminar flow hood, biosafety cabinet, cleanroom, or inside an isolator. The bottle holder is ergonomically positioned around the pump head for easy access in limited-space environments. The bottle holder with the fastening screws is used for slim bottles. The height and orientation of the bottle holder basket can be adjusted according to user requirements.

Application

The Steritest® Symbio Slim Bottle Holder is an accessory for the Steritest® Symbio pumps for streamlining the workflow and increasing safety during sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service