Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SX1244

Supelco

Sulfuric acid

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Oil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-639-5

Agency

meets USP/NF

Quality Level

vapor density

<0.3 (25 °C, vs air)

vapor pressure

0.0001 hPa ( 20 °C)
1 mmHg ( 146 °C)

Assay

95.0-98.0%

form

liquid

impurities

≤2 ppm Substances reducing permanganate (as SO2)

ign. residue

≤5 ppm

color

APHA: ≤10

pH

0.3 (25 °C, 49 g/L in H2O)

bp

~290 °C (lit.)

mp

-20 °C

density

1.840 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.2 ppm
nitrate (NO3-): ≤0.5 ppm

cation traces

Al: ≤0.2 ppm
As: ≤0.01 ppm
Au: ≤0.3 ppm
Ca: ≤0.3 ppm
Cr: ≤0.2 ppm
Cu: ≤0.1 ppm
Fe: ≤0.2 ppm
Hg: ≤5 ppb
K: ≤0.3 ppm
Mg: ≤0.3 ppm
Mn: ≤0.2 ppm
NH4+: ≤2 ppm
Na: ≤0.3 ppm
Ni: ≤0.1 ppm
Pb: ≤0.3 ppm
Sn: ≤0.2 ppm
Ti: ≤0.3 ppm
Zn: ≤0.2 ppm
heavy metals (as Pb): ≤1 ppm

shipped in

ambient

storage temp.

room temp

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
SX1244PC-5 and SX1244-75 are packaged in poly coated glass bottles

Analysis Note

Aluminum (Al): 0.2ppm max
Ammonium (NH4): 2ppm max
Appearance: To pass test
Arsenic (As): 0.01ppm max
Assay: 95.0-98.0%
Calcium (Ca): 0.3ppm max
Chloride (Cl): 0.2ppm max
Chromium (Cr): 0.2ppm max
Color (APHA): 10 max
Copper (Cu): 0.1ppm max
Gold (Au): 0.3ppm max
Heavy metals (as Pb): 1ppm max
Iron (Fe): 0.2ppm max
Lead (Pb): 0.3ppm max
Magnesium (Mg): 0.3ppm max
Manganese (Mn): 0.2ppm max
Mercury (Hg): 5ppb max
Nickel (Ni): 0.1ppm max
Nitrate (NO3): 0.5ppm max
Potassium (K): 0.3ppm max
Residue after ignition: 5ppm max
Sodium (Na): 0.3ppm max
Substances reducing permanganate (as SO2): 2ppm max
Tin (Sn): 0.2ppm max
Titanium (Ti): 0.3ppm max
Zinc (Zn): 0.2ppm max

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service