Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SCT260

Sigma-Aldrich

BioTracker Green Lipid Peroxide Live Cell Ferroptosis Indicator

Human

Synonym(s):

lipid peroxide live cell dye, lipid peroxide probe, live cell ferroptosis indicator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.47

Product Name

BioTracker Green Lipid Peroxide Live Cell Ferroptosis Indicator,

biological source

human

Quality Level

packaging

vial of 250 μg

manufacturer/tradename

Millipore

technique(s)

cell culture | stem cell: suitable
flow cytometry: suitable

shipped in

2-8°C

storage temp.

2-8°C

General description

This perylene derivative probe containing oligooxyethylene is designed for detection of lipid peroxides. BioTracker Green Lipid Peroxide Live Cell Ferroptosis Indicator emits intense fluorescence by lipid peroxide specific oxidation in organic solvents such as ethanol. This is the only RUO probe that can specifically detect lipid peroxides.SCT260′s oxidized form is nearly nonfluorescent in an aqueous media, but produces a bright fluorescence in lipophilic sites such as in cell membranes.This probe is light-sensitive, and should be pipetted as quickly as possible, preferably within one minute. Protect working solutions and samples from light, and use within one day of preparing working solution.

Features and Benefits

This perylene derivative probe containing oligooxyethylene is designed for the detection of lipid peroxides in live cells.

Target description

This perylene derivative containing oligooxyethylene is designed for the detection of lipid peroxides. SCT260 emits intense fluorescence by lipid peroxide-specific oxidation in organic solvents such as ethanol. Among fluorescent probes that detect Reactive Oxygen Species(ROS), SCT260 is the only compound that can specifically detect lipid peroxides. Because the excitation and emission wavelengths of the oxidized probe are 524 nm and 535 nm, respectively, phototoxicity to and autofluorescence from the sample can be minimized. The tetraethyleneglycol group linked to one end of diisoquinoline ring enhances solubility and dispersibility in aqueous buffer.

Storage and Stability

Store BioTracker Green Lipid Peroxide Live Cell Ferroptosis Indicator at 2 - 8 °C, protected from light.

Other Notes

Live cell probe for fluorescent detection of lipid peroxidation by fluorescence microscopy or flow cytometry. SCT260 is a valuable reagent for ferroptosis research.

Legal Information

BioTracker is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service