Skip to Content

SCT019

BioTracker 560 Orange Lysosome Dye

Live cell imaging dye for acidic cellular organelles such as lysosomes.

Synonym(s):

Live cell imaging probe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.47
eCl@ss:
32161000
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

ambient

storage temp.

-10 to -25°C

General description

Lysosomes are membrane-enclosed organelles that contain an array of enzymes capable of breaking down all types of biological material including proteins, nucleic acids, carbohydrates, and lipids. Lysosomes function as the digestive system of the cell, serving both to degrade material taken up from outside the cell and to digest obsolete components of the cell itself.

The BioTracker 560 Orange Lysosome Dye is a live cell fluorescence imaging probe which enhances fluorescence dramatically in acidic environments. This probe can stain acidic organelles such as lysosome, late endosome and granule selectively. Namely, in the condition of pH 5.0, identical to the environment of acidic organelles, fluorescence intensity is 50-fold or more compared to physical pH 7.4. The BioTracker 560 Orange Lysosome Dye shows great photostability against irradiating excitation light. As it emits orange color fluorescent by excitation at 532nm or 514 nm, multicolor imaging is possible by combining with blue fluorescence (CFP, Hoechst, etc.), green fluorescence (GFP, fluorescein, etc.) and near-infrared fluorescence.

Spectral Properties
Absorbance: 535nm
Emission: 560nm

Application

Live cell fluorescent imaging
Research Category
Cell Imaging
Research Sub Category
Live Cell Dye

Physical form

Lyophilized

Preparation Note

Store BioTracker 560 Orange Lysosome Dye at -20°C, desiccate and protect from light
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library