Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCMF001

Sigma-Aldrich

FIBROGRO® Complete Media Kit for Culture of Human Fibroblasts

The FibroGRO Media provide an optimal cell culture environment for many types of human fibroblasts & mouse embryonic fibroblasts.

Synonym(s):

FibroGRO-LS Medium, Human Fibroblast Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

Quality Level

product line

Chemicon®

form

liquid

manufacturer/tradename

Chemicon®

storage condition

do not freeze

technique(s)

cell culture | mammalian: suitable

input

sample type epithelial cells

General description

FibroGRO® Media formulations provide an optimal cell culture environment for many types of human fibroblasts and mouse embryonic fibroblasts These low-serum/serum-free formulations have been shown to grow these cells at rates that meet or exceed commercially available serum containing media, while maintaining excellent cell morphology. The media are packaged in a specially designed UV protective shrinkwrap for added stability, which includes a temperature gauge for added convenience. FibroGRO® Media contain no antimicrobials and no phenol red, components that can cause cell stress and masking effects that may influence experimental results.

Application

FibroGRO Complete Medium is a defined medium optimized for serum-free culture of human fibroblasts, and supports the growth of these cells in a serum-free environment at rates equal to or greater than comparable media supplemented with much higher concent
FibroGRO Complete Medium is a defined medium optimized for serum-free culture of human fibroblasts, and supports the growth of these cells in a serum-free environment at rates equal to or greater than comparable media supplemented with much higher concentrations of FBS. FibroGRO Medium contains no antimicrobials and no phenol red; components that can cause cell stress and “masking effects” that may influence experimental results (these are not needed, or recommended, to achieve optimal cell performance). FibroGRO medium provides a perfect cell system to establish serum-free human feeder layers for stem cell culture, or as a model to study wound healing, toxicology or basic cell biology
The FibroGRO Media provide an optimal cell culture environment for many types of human fibroblasts & mouse embryonic fibroblasts.

Components

  • SCMF-BM
  • SCMF001-S

Storage and Stability

SCMF-BM: Store at 4°C. See label for expiry date. SCMF001-S: Store at -20°C. See individual labels for expiry date. Supplemented medium = 4°C, in dark. Use within 1 month of supplementation. Do not freeze.

Other Notes

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
FIBROGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service