Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

SCC261

Sigma-Aldrich

Oli-neu Mouse Oligodendroglial Precursor Cell Line

Synonym(s):

OLIneu, oliNeu, olineu

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.81

packaging

vial of ≥1X10⁶ cells/vial mg

Quality Level

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

cell culture | stem cell: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

General description

Oli-neu is an immortalized mouse oligodendroglial cell line that recapitulates morphology and in vivo physiological activity of oligodendroglial cells. Oli-neu cells differentiate into oligodendrocytes upon daily addition of 1 mM dibutyrl-cAMP.2 Oli-neu cells have been utilized in a broad range of studies of oligodendrocyte biology and has greatly contributed to the understanding of the mechanisms of myelination and molecular players in myelination disorders. Differentiated Oli-neu oligodendrocytes express O4 antigen, an oligodendrocyte marker, and manifest cell processes characteristic of oligodendrocyte morphology.2 The Oli-neu cell line is a versatile cellular model for oligodendrocyte biology and associated neurophysiological investigations.

Application

  • Each vial contains > 1X106 viable cells.
  • Cells are tested negative for infectious diseases by a Mouse Essential CLEAR Panel by Charles River Animal Diagnostic Services.
  • Cells are verified to be of mouse origin and negative for inter-species contamination from rat, human, chinese hamster, Golden Syrian hamster, and Non-human Primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
  • Cells are negative for mycoplasma contamination.

Oligodendrocytes, comprising 5-10% of the total population of glial cells in the central nervous system, are responsible for producing the myelin sheath encasing axons which is essential for efficient electric conduction of neural impulses.1 Oligodendrocytes arise from precursor cells that differentiate just before birth. Disorders of myelination of axons can lead to emergence of neurodegenerative diseases such as Alzheimer′s and multiple sclerosis, underscoring the critical importance of understanding oligodendrocyte biology and interactions. The myelination process occurs within the complex milieu of neuronal cell types, thus is difficult to reconstruct without cellular models. Oli-neu is an immortalized mouse oligodendroglial cell line that recapitulates morphology and in vivo physiological activity of oligodendroglial cells. Oli-neu cells differentiate into oligodendrocytes upon daily addition of 1 mM dibutyrl-cAMP.2 Oli-neu cells have been utilized in a broad range of studies of oligodendrocyte biology and has greatly contributed to the understanding of the mechanisms of myelination and molecular players in myelination disorders. Differentiated Oli-neu oligodendrocytes express O4 antigen, an oligodendrocyte marker, and manifest cell processes characteristic of oligodendrocyte morphology.2 The Oli-neu cell line is a versatile cellular model for oligodendrocyte biology and associated neurophysiological investigations.Source Oli-neu was derived from oligodendrocytes and oligodendrocyte precursor cells isolated from the cerebellum of a 6-day-old NMRI mouse and infected with retrovirus containing the t-neu tyrosine kinase oncogene.2References1. Curr Pharm Des. 2016, 22(6): 656-679.2. Eur J Neurosci. 1995, 7(6): 1245-1265.

Features and Benefits

The Oli-neu cell line is a versatile cellular model for oligodendrocyte biology and associated neurophysiological investigations.

Storage and Stability

Oli-neu mouse oligodendrocyte precursor cells should be stored in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting functionality.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service