Skip to Content
MilliporeSigma
All Photos(1)

Documents

S8001

Sigma-Aldrich

CpGenome Human Methylated & Non-Methylated DNA Standard Set

Synonym(s):

DNA Standard Set, Methylated DNA Standard Set, Non-Methylated DNA Standards

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106310
NACRES:
NA.84

Quality Level

General description

CpGenome Human Methylated & Non-methylated DNA Standards are purified from HCT116 DKO cells that contain genetic knockouts of DNA methyltransferases, DNMT1 (-/-) and DNMT3b (-/-) , and have less than 5% methylated DNA. The CpGenome Human Methylated DNA Standard has been methylated enzymatically at all CpG dinucleotides by M. SssI methyltransferase. These standards are intended for use as positive and negative controls in gene methylation studies, such as bisulfite conversion of DNA with the CpGenome Turbo Bisulfite Modification Kit (Cat. No. S7847). Bisulfite-modified DNA can be evaluated by methylation-specific PCR (MSP) technology using the CpG WIZ amplification kits.

Materials Provided:
Two vials, each containing 5 μg (20 μl) of CpGenome Human Non-Methylated DNA Standard or 5 μg (20 μl) of CpGenome Human Methylated DNA Standard, at a concentration of 250 ng/μL.

Validation:
Methylation-specific PCR (MSP) was performed on the DNA after bisulfite modification with the CpGenome Turbo Bisulfite kit (Cat. No. S7847). Three sets of primers from the CpG WIZ BRCA1 amplification kit (Cat. No. S7830) were used in this assay: the U primer set, which anneals to unmethylated bisulfite-modified DNA; M primers, which anneal to methylated bisulfite-modified sequences; and W primers, which anneal to unmethylated or methylated DNA that has not undergone bisulfite modification. Only the M primers generated product for the methylated DNA standard, and only the U primer set amplified the unmethylated Standard.

CpGenome and CpG WIZ are trademarks of Serologicals Corporation. CpG WIZ Methylation Products apply to technologies exclusively licensed from The Johns Hopkins University School of Medicine. Methylation-specific PCR (MSP) technology is covered under U.S. Patent # 5,786,146.

Application

Purified from HCT116 DKO cells that contain genetic knockouts of DNA methyltransferases, DNMT1 (-/-) & DNMT3b (-/-) & have <5% methylated DNA.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service