Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8570

Sigma-Aldrich

Methylated Control DNA

50 ng/μL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
NA.55

form

liquid

concentration

50 ng/μL

color

colorless

storage temp.

−20°C

General description

Fully methylated Jurkat DNA. 5-methyl-cytidine (5mC) content determined by mass spectrometry. See lot specific certificate of analysis or vial label for 5mC content of each lot.

Application

Methylated Control DNA has been used as a negative control in bisulfite sequencing of embryonic genomic DNA.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Non-CpG methylation by DNMT3B facilitates REST binding and gene silencing in developing mouse hearts
Zhang D, et al.
Nucleic Acids Research, 45(6), 3102-3115 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service