Skip to Content
MilliporeSigma
All Photos(2)

Documents

S2VPU11RE

Millipore

Millipore® Stericup® Quick Release Vacuum Filtration System

pore size 0.1 μm, polyethersulfone membrane, funnel capacity 1000 mL, pack of 12 ea

Synonym(s):

SCVPU11RE, Stericup Quick Release-VP Sterile Vacuum Filtration System, sterile vacuum filter unit, sterile vacuum filtering unit, vacuum filtration system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104914
NACRES:
NB.24

material

polyethersulfone membrane
polystyrene bottle
polystyrene funnel(s)
polystyrene housing

Quality Level

sterility

sterile; irradiated

product line

Stericup®

feature

hydrophilic

packaging

pack of 12 ea

parameter

max. temp. at 45 °C

description

2.4 mL (hold-up volume)

technique(s)

sterile filtration: suitable

H

319 mm

capacity

1000 mL

diam.

13.5 cm

filter diam.

73 mm

filtration area

40 cm2

funnel capacity

1000 mL

matrix

Durapore®

pore size

0.1 μm pore size

fitting

inlet funnel
outlet flask (Receiver)

shipped in

ambient

General description

This item is a direct replacement for SCVPU11RE
Work with ease and filter with confidence with Stericup® and Steritop® Filtration Systems.

Stericup® and Steritop® sterile filtration devices use reliable, high-performance membranes for optimum sterile filtration of laboratory reagents, buffers, and media. These devices combine superior flow rates and throughput with low non-specific binding and a stable, no-tip design.

This Stericup® 0.1 μm pore size polyethersulfone (PES) membrane filter unit combines a 1000 mL funnel with a 1000 mL receiver flask and cap for processing and storage.

Application

For Research Use Only
Stericup® and Steritop® systems are designed to reduce foaming and protein denaturation while maximizing flow and can be used for sterile vacuum filtration of aqueous solutions such as tissue culture media and biological fluids.

  • Tissue culture media +/- additives
  • Buffers
  • Biological solutions

Features and Benefits

  • The Quick Release filter funnel disconnects from the receiver bottle with just a quarter turn—reducing the likelihood of spillage and making it easier to manipulate on the bench or under the hood.
  • Bold product identification on the Steritop® filter funnel plainly displays membrane characteristics, including composition and pore size, for rapid, application-appropriate product selection.
  • Click seal cap is flanged and modified to enable a firm grip in wet or dry conditions—and a tactile stop confirms secure closure. Worry less about the possibility of spilling or contamination of sterilized contents.
  • Frosted, ample writing surface on the receiver bottle and lighter cap color facilitate clear labelling to improve legibility and reduce unreadable smears.
  • Click seal cap rests on the side to avoid risk of contamination.
  • Receiver bottles are stackable to save storage space.

Legal Information

Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Stericup is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Evaluation of mycoplasma removal by sterilizing filtration using 0.1 µm PES and PVDF filters for cell culture media and buffer preparation.

Evaluation of mycoplasma removal by sterilizing filtration using 0.1 µm PES and PVDF filters for cell culture media and buffer preparation.

Evaluation of mycoplasma removal by sterilizing filtration using 0.1 µm PES and PVDF filters for cell culture media and buffer preparation.

Evaluation of mycoplasma removal by sterilizing filtration using 0.1 µm PES and PVDF filters for cell culture media and buffer preparation.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service