Skip to Content
MilliporeSigma

Skip To

S2GPU10RE

Millipore® Stericup® Quick Release Vacuum Filtration System

pore size 0.22 μm, polyethersulfone membrane, filter capacity 500 mL, pack of 12 ea

Synonym(s):

SCGPU10RE

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41104914
NACRES:
NB.24
eCl@ss:
32031623
Pore size:
0.22 μm pore size
Filter capacity:
500 mL
Packaging:
pack of 12 ea
Material:
polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing
Sterility:
sterile
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

sterility

sterile

product line

Stericup®

feature

hydrophilic

packaging

pack of 12 ea

parameter

45 °C max. temp.

description

2.4 mL (hold-up volume)

technique(s)

sterile filtration: suitable

H

299 mm

diam.

12.5 cm

filter capacity

500 mL

filter diam.

73 mm

filtration area

40 cm2

matrix

Express PLUS

pore size

0.22 μm pore size

fitting

inlet funnel, outlet flask (Receiver)

shipped in

ambient

storage temp.

no temp limit

General description

This item is a direct replacement for SCGPU10RE
Work with ease and filter with confidence with Stericup® and Steritop® Filtration Systems.

Stericup® and Steritop® sterile filtration devices use reliable, high-performance membranes for optimum sterile filtration of laboratory reagents, buffers, and media. These devices combine superior flow rates and throughput with low non-specific binding and a stable, no-tip design.

This Stericup® 0.22 μm pore size polyethersulfone (PES) membrane filter unit combines a 500 mL bottle-top with a 1000 mL receiver flask and cap for processing and storage.

Application

For Research Use Only
Stericup® and Steritop® systems are designed to reduce foaming and protein denaturation while maximizing flow and can be used for sterile vacuum filtration of aqueous solutions such as tissue culture media and biological fluids.

  • Tissue culture media +/- additives
  • Buffers
  • Biological solutions

Features and Benefits

  • The Quick Release filter funnel disconnects from the receiver bottle with just a quarter turn—reducing the likelihood of spillage and making it easier to manipulate on the bench or under the hood.
  • Bold product identification on the Steritop® filter funnel plainly displays membrane characteristics, including composition and pore size, for rapid, application-appropriate product selection.
  • Click seal cap is flanged and modified to enable a firm grip in wet or dry conditions—and a tactile stop confirms secure closure. Worry less about the possibility of spilling or contamination of sterilized contents.
  • Frosted, ample writing surface on the receiver bottle and lighter cap color facilitate clear labelling to improve legibility and reduce unreadable smears.
  • Click seal cap rests on the side to avoid risk of contamination.
  • Receiver bottles are stackable to save storage space.

Legal Information

EXPRESS PLUS is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Stericup is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S2GPU11RES2GPU01RES2GPU02RE
sterility

sterile

sterility

sterile

sterility

-

sterility

sterile

filter capacity

500 mL

filter capacity

1000 mL

filter capacity

150 mL

filter capacity

250 mL

material

polyethersulfone membrane, polystyrene funnel(s), polystyrene bottle, polystyrene housing

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

pore size

0.22 μm pore size

pore size

0.22 μm pore size

pore size

0.22 μm pore size

pore size

0.22 μm pore size

packaging

pack of 12 ea

packaging

pack of 12 ea

packaging

pack of 12 ea

packaging

pack of 12 ea

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic


Still not finding the right product?




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–10 of 15 Questions  
  1. What is the difference between classic Stericup® filters and Stericup® “Quick Release” filters?

    1 answer
    1. Stericup® Quick Release, or “QR”, filters feature a connection between funnel filter and receiver bottle that requires only a quarter-turn release to facilitate funnel removal. These filters come fully engaged – there is no need to tighten before use. The receiver bottle cap has flanges on the top surface to improve grip, as well as a positive lock stop to ensure a leak-proof seal with the receiver bottle. Other improvements include a large, frosted writing surface on the receiver bottle and a lighter cap color to facilitate labelling, more prominent volume markings on the filtration funnel and receiver bottle, and a product label with a peel-off lot record strip on the bag for easy viewing in storage.

      Helpful?

  2. Can we filter oil-type liquids with Stericup® filters?

    Can we filter oil-type liquids with Stericup® filters?

    1 answer
    1. Likely not, due to pore size/possible fouling/chemical compatibility. Refer to our guide, "Chemical compatibility of filter components": https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/302/855/membrane-chemical-compatibility-ds2532en-ms.pdf

      Helpful?

  3. Which type of pump should be used with Stericup® and Steritop® vacuum filters?

    Which type of pump should be used with Stericup® and Steritop® vacuum filters?

    1 answer
    1. We recommend our Chemical Duty Pump: https://www.sigmaaldrich.com/product/mm/wp6111560

      Helpful?

  4. What Bacterial Endotoxins Test is used?

    1 answer
    1. The test method used complies with USP 85 and EP 2.6.14 to ensure the absence of endotoxin in our products.

      Helpful?

  5. Are Stericup® and Steritop® filters GMP-certified?

    1 answer
    1. No, Stericup® and Steritop® devices are not GMP-approved.

      Helpful?

  6. What tubing can be used with Stericup® and Steritop® vacuum filters?

    What tubing can be used with Stericup® and Steritop® vacuum filters?

    1 answer
    1. Stericup® and Steritop® filters are fitted with a 1/4” tube adapter. We recommend 3/16 in. (4.8 mm) ID silicone tubing to fit this tube adapter: https://www.sigmaaldrich.com/product/mm/xx7100004

      Helpful?

  7. What is the maximum vacuum pressure for filtration?

    1 answer
    1. 700 mm Hg (27.5” Hg, 0.9 bar) differential vacuum at 25 °C (77 °F)

      Helpful?

  8. Can Stericup® and Steritop® filters be reused?

    1 answer
    1. All Stericup® and Steritop® filters were developed for single use, disposable processing.

      Helpful?

  9. What leachables are detected during filtration processes using Stericup® filters?

    1 answer
    1. We have not performed extractables/leachables studies for our Stericup® filters.

      Helpful?

  10. Can Stericup® and Steritop® filters be used to prepare solutions that will be administered to patients?

    1 answer
    1. No, all Stericup® and Steritop® devices are Research Use Only.

      Helpful?

1–10 of 15 Questions  

Reviews

No rating value

Active Filters