Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PX2020

Supelco

Pyridine

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Pyridine, Azabenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N
CAS Number:
Molecular Weight:
79.10
MDL number:
UNSPSC Code:
12191501
EC Index Number:
203-809-9
Assay:
≥99% (GC)
bp:
115 °C/1013 hPa
vapor pressure:
26.7 hPa ( 25 °C)
Pricing and availability is not currently available.

Agency

meets USP/NF

Quality Level

vapor pressure

26.7 hPa ( 25 °C)

Assay

≥99% (GC)

form

liquid

impurities

≤0.001% Chloride (Cl)
≤0.001% Sulfate (SO4)
≤0.002% Ammonia (NH3)
≤0.1% Water (H2O)
≤5 ppm Copper (Cu)

evapn. residue

≤0.002%

pH

8.81 (20 °C in H2O)

bp

115 °C/1013 hPa

mp

-42 °C

transition temp

flash point 17 °C

density

0.982 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

SMILES string

n1ccccc1

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H

InChI key

JUJWROOIHBZHMG-UHFFFAOYSA-N

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Ammonia (NH3): 0.002% max
Assay (GC): 99.0% min
Chloride (Cl): 0.001% max
Copper (Cu): 5ppm max
Evaporation residue: 0.002% max
Form: Clear liquid
Reducing substances: To pass test
Solubility in water: To pass test
Sulfate (SO4): 0.001% max
Water (H2O): 0.1% max
Evaporation Number: 12.7

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service