Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.94601

Supelco

Pyridine

for analysis EMPARTA® ACS

Synonym(s):

Pyridine, Azine, Azabenzene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
79.1
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-809-9
NACRES:
NB.21

grade

ACS reagent

vapor pressure

26.7 hPa ( 25 °C)

product line

EMPARTA®

Assay

≥99% (GC)

form

liquid

autoignition temp.

482 °C

potency

891 mg/kg LD50, oral (Rat)
1121 mg/kg LD50, skin (Rabbit)

pH

8.81 (20 °C in H2O)

bp

115 °C/1013 hPa

mp

-42 °C

transition temp

flash point 17 °C

solubility

soluble 1000 g/L

density

0.982 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H

InChI key

JUJWROOIHBZHMG-UHFFFAOYSA-N

General description

for analysis EMPARTA® ACS

Application


  • Vinylcyclopropane-Cyclopentene (VCP-CP) Rearrangement Enabled by Pyridine-Assisted Boronyl Radical Catalysis.: This study explores the VCP-CP rearrangement facilitated by pyridine-assisted boronyl radical catalysis, highlighting its implications for synthetic chemistry and potential applications in creating complex molecular structures efficiently (Li et al., 2024).

Analysis Note

Evaporation Number: 12.7

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service