Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PX1838

Supelco

2-Propanol

≥99.8% (GC), suitable for HPLC, suitable for SM 4500 - NH3, meets USP/NF, suitable for SM 2320, suitable for SM 2310

Synonym(s):

2-Propanol, 2-Propanol,IPA, 2-Propanol, sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-661-7
Assay:
≥99.8% (GC)
technique(s):
HPLC: suitable
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
43 hPa ( 20 °C)
44 mmHg ( 25 °C)
Pricing and availability is not currently available.

Product Name

Isopropyl Alcohol, HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Agency

meets USP/NF
suitable for SM 2310
suitable for SM 2320
suitable for SM 4500 - NH3

Quality Level

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
43 hPa ( 20 °C)
44 mmHg ( 25 °C)

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

technique(s)

HPLC: suitable

impurities

≤0.0001 meq/g Titrable acid
≤0.0001 meq/g Titrable base
≤0.002% Carbonyl compounds (acetone or propionaldehyde)
≤0.10% Water (H2O)

evapn. residue

≤5 ppm

color

APHA: ≤10

refractive index

n20/D 1.377 (lit.)

pH

7 (20 °C)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

transition temp

flash point 12 °C

density

0.785 g/mL at 25 °C (lit.)

UV cutoff

≤210 nm

UV absorption

λ: 210 nm Amax: ≤1.00
λ: 220 nm Amax: ≤0.40
λ: 230 nm Amax: ≤0.20
λ: 245 nm Amax: ≤0.08
λ: 260 nm Amax: ≤0.04
λ: 275 nm Amax: ≤0.03
λ: 300 nm Amax: ≤0.02
λ: 330-400 nm Amax: ≤0.01

shipped in

ambient

storage temp.

room temp

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
PX1838P-6 andPX1838P-1 are packaged in poly coated glass bottles

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

AppearanceClear liquid, free from particulatesAssay (GC)99.8% minCarbonyl compounds (acetone or propionaldehyde)0.002% maxColor (APHA)10 maxEvaporation residue5ppm maxFiltered through 0.2 µm filterTo pass testInfrared SpectrumConforms to standardSolubility in waterTo pass testTitrable acid0.0001meq/g maxTitrable base0.0001meq/g maxUV Abs. at 210 nm1.00AU maxUV Abs. at 220 nm0.40AU maxUV Abs. at 230 nm0.20AU maxUV Abs. at 245 nm0.08AU maxUV Abs. at 260 nm0.04AU maxUV Abs. at 275 nm0.03AU maxUV Abs. at 300 nm0.02AU maxUV Abs. at 330 - 400 nm0.01AU maxUV Cut-off210nm maxWater (H2O)0.10% max

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service