Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PX1835

Supelco

2-Propanol

≥99.5% (GC), meets USP/NF

Synonym(s):

2-Propanol, 2-Propanol,IPA, 2-Propanol, sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-661-7
Assay:
≥99.5% (GC)
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
43 hPa ( 20 °C)
44 mmHg ( 25 °C)
Pricing and availability is not currently available.

Product Name

Isopropyl Alcohol, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Agency

meets USP/NF

Quality Level

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
43 hPa ( 20 °C)
44 mmHg ( 25 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

impurities

≤0.0001 meq/g Titrable acid
≤0.0001 meq/g Titrable base
≤0.002% Carbonyl compounds (acetone or propionaldehyde)
≤0.2% Water (H2O)
≤2 ppm Benzene (GC)

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.377 (lit.)

pH

7 (20 °C)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

transition temp

flash point 12 °C

density

0.785 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
PX1835P-4 is packaged in poly coated glass bottles

Analysis Note

Assay (GC): 99.5% min
Carbonyl compounds (acetone or propionaldehyde): 0.002% max
Color (APHA): 10 max
Evaporation residue: 0.001% max
Form: Clear liquid
Infrared Spectrum: Conforms to standard
Solubility in water: To pass test
Titrable acid: 0.0001meq/g max
Titrable base: 0.0001meq/g max
Water (H2O): 0.2% max

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Is 2-propanol (IPA) available in a plastic bottle?

    1 answer
    1. The products PX1835-9 and PX1835-7 are both available in plastic bottles.

      Helpful?

  2. What is the storage temperature range for item PX1835, Isopropyl Alcohol?

    1 answer
    1. The recommended storage temperature for item PX1835, Isopropyl Alcohol, is between +2°C to +25°C

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service