Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PX1565

Supelco

Potassium phosphate monobasic

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Potassium Dihydrogen Phosphate, Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-913-4
Assay:
≥99% (acidimetric)
form:
crystals
solubility:
soluble 208 g/L

Agency

meets USP/NF

Quality Level

Assay

≥99% (acidimetric)

form

crystals

impurities

≤0.01% Insoluble matter

loss

≤ 0.2% loss on drying, 105°C

color

white

pH

4.2-4.6 (20 °C, 20 g/L in H2O)

mp

252.6 °C (lit.)

solubility

soluble 208 g/L

density

2.338 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.003%

cation traces

Fe: ≤0.002%
Na: ≤0.005%
heavy metals: ≤0.001%

shipped in

ambient

storage temp.

room temp

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay (acidimetric)99.0% minChloride (Cl)0.001% maxColorWhiteFormCrystalsHeavy Metals (by ICP-OES)0.001% maxInsoluble matter0.01% maxIron (Fe)0.002% maxLoss on drying (105°C)0.2% maxSodium (Na)0.005% maxSulfate (SO4)0.003% maxpH of a 5% solution at 25°C4.1-4.5

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service