Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PX0167

Supelco

Pentane

OmniSolv®

Synonym(s):

high purity solvents

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-692-4
technique(s):
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
bp:
35-36 °C (lit.)
vapor pressure:
570 hPa ( 20 °C)

vapor density

2.48 (vs air)

Quality Level

vapor pressure

570 hPa ( 20 °C)

product line

OmniSolv®

Assay

≥98% n-pentane basis
≥99.9% (C5 isomers)

form

liquid

autoignition temp.

500 °F

expl. lim.

~8.3 %

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.01% Water (H2O)
≤10 ng/L Capillary ECD responsive substances (as PCNB)

evapn. residue

≤1 mg/L

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

-130 °C
−130 °C (lit.)

transition temp

flash point -48 °C (refers to pure substance)

density

~0.63 g/cm3 at 20 °C
0.626 g/mL at 25 °C (lit.)

UV absorption

λ: 190 nm Amax: ≤1.00
λ: 200 nm Amax: ≤0.600
λ: 250 nm Amax: ≤0.010
λ: 300 nm Amax: ≤0.005
λ: 400 nm Amax: ≤0.005

shipped in

ambient

storage temp.

15-25°C

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

OmniSolv®

Analysis Note

Assay (n-Pentane)98% minAssay (total C5 isomers)99.9% minCapillary ECD responsive substances (as PCNB)10ng/L maxEvaporation residue1mg/L maxFormClear liquidRefractive index (at 20°C)1.3570-1.3580UV Abs. at 190 nm1.00AU maxUV Abs. at 200 nm0.600AU maxUV Abs. at 250 nm0.010AU maxUV Abs. at 300 nm0.005AU maxUV Abs. at 400 nm0.005AU maxWater (H2O)0.01% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service