Skip to Content
MilliporeSigma
All Photos(1)

Documents

PRPK00001

Milli-Q

PrePak Pretreatment Pack

For use with Elix®, AFS® and RiOs systems, Protects and maintains good performance of the reverse osmosis (RO) membrane

Synonym(s):

PrePak 1 Pretreatment Pack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
JA.13

Quality Level

feature

Optimal protection of water system and optimised performance; activated C; SDI > 5 & < 12

packaging

pkg of 2 units

pore size

0.5 μm pore size (CRW5 type)

compatibility

for use with AFS®
for use with Elix®
for use with RiOs

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Proper pretreatment is an essential step in producing the desired final product water quality. The choice of the pretreatment is made according to the flow rate of the feed water.
The PrePak 1 pretreats tap water by removing particles, organic compounds, and free chlorine. It is intended for use with larger water purification systems as additional pretreatment to the system′s Progard® TL pretreatment packs.
For use with Elix®, AFS®, and RiOs water purification systems.

Application

Protects and maintains good performance of the reverse osmosis (RO) membrane.

Features and Benefits

  • Optimal protection of the water system.
  • Ensures system lifetime and performance are optimized.
  • 0.5 μm depth-filtration to remove particles.
  • Activated carbon to remove chlorine.

Legal Information

AFS is a registered trademark of Merck KGaA, Darmstadt, Germany
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
PROGARD is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service