Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHCC3CHARG

Millipore

Scepter 3.0 Charging Station

Synonym(s):

3.0 Charging Dock, Scepter Charging Station, Station for Charging Scepter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41131501
NACRES:
NC.57

packaging

pkg of 1 ea

manufacturer/tradename

Scepter

technique(s)

cell counting: suitable

compatibility

Scepter

shipped in

ambient

General description

The Scepter 3.0 Charging Station can be mounted (using the Scepter 3.0 Charging Station Mounting Kit, sold separately) in multiple mounting options or can be stored stationary on the bench. The Charging Station stores Scepter 3.0 out-of-the-way when not in use, freeing up valuable hood or lab bench space.

A Charging Station is included with Scepter 3.0 instruments and can also be purchased separately for mulitple charging options.

Components

Included with Scepter 3.0 instruments:PHCC340KITPHCC360KITCan be purchased as separately for additional charging stations for Scepter 3.0

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The Scepter™ 3.0 Cell Counter, when used in conjunction with a 40 μm aperture sensor, can accurately count and precisely size a wide range of cell types and particles. This application note demonstrates two examples of experiments using the instrument’s sensitive size-discriminating capacity for rapid qualitative assessment of individual cell populations.

The Scepter™ 3.0 Cell Counter, when used in conjunction with a 40 μm aperture sensor, can accurately count and precisely size a wide range of cell types and particles. This application note demonstrates two examples of experiments using the instrument’s sensitive size-discriminating capacity for rapid qualitative assessment of individual cell populations.

The Scepter™ 3.0 Cell Counter, when used in conjunction with a 40 μm aperture sensor, can accurately count and precisely size a wide range of cell types and particles. This application note demonstrates two examples of experiments using the instrument’s sensitive size-discriminating capacity for rapid qualitative assessment of individual cell populations.

The Scepter™ 3.0 Cell Counter, when used in conjunction with a 40 μm aperture sensor, can accurately count and precisely size a wide range of cell types and particles. This application note demonstrates two examples of experiments using the instrument’s sensitive size-discriminating capacity for rapid qualitative assessment of individual cell populations.

Related Content

Scepter™ Cell Counter is a portable device which brings consistency in cell counting right to the culture hood in less than 30 seconds.

Scepter™ Cell Counter is a portable device which brings consistency in cell counting right to the culture hood in less than 30 seconds.

Scepter™ Cell Counter is a portable device which brings consistency in cell counting right to the culture hood in less than 30 seconds.

Scepter™ Cell Counter is a portable device which brings consistency in cell counting right to the culture hood in less than 30 seconds.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service